Learn More
Silver iodide, 99%, pure
Product Code. 10370984 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10370984 100 g Glass bottle 100g
10072790 5 g Glass bottle 5g
10490481 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 10370984 Supplier Thermo Scientific Acros Supplier No. 210421000
Request Bulk or Custom Format
CAS: 7783-96-2 | AgI | 234.77 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 7783-96-2
Molecular Formula AgI
Molecular Weight (g/mol) 234.77
MDL Number MFCD00003412
InChI Key MSFPLIAKTHOCQP-UHFFFAOYSA-M
Synonym silver iodide, neosiluol, neosilvol, silver monoiodide, silver 1+ iodide, colloidal silver iodide, iodanylsilver, silver iodide metals basis, ccris 8490
PubChem CID 24563
IUPAC Name iodosilver
SMILES [Ag]I
Specifications
Boiling Point 1506°C
Melting Point 557°C
Color Yellow
Physical Form Granular Powder
Quantity 100 g
Assay Percent Range 98.5% min. (ex Ag)
Linear Formula AgI
Merck Index 15, 8655
Solubility Information Solubility in water: 0.03mg/l
Formula Weight 234.77
Percent Purity 99%
Grade Pure
Packaging Glass bottle
Specific Gravity 5.68
Density 5.6800g/mL
Chemical Name or Material Silver iodide, Pure
Show More Show Less
compliance-icons
Product Identifier
  • Silver iodide
Signal Word
  • Warning
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
Hazard Statement
  • H410-Very toxic to aquatic life with long lasting effects.
  • H400-Very toxic to aquatic life.
Precautionary Statement
  • P273-Avoid release to the environment.