missing translation for 'onlineSavingsMsg'
Learn More

Silver Nitrate, 0.1N Volumetric Solution, BAKER ANALYZED™ Reagent, J.T. Baker™

Product Code. 15953077 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
1 L
Packaging:
Narrow Mouth Amber Glass Bottle
Unit Size:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15953077 1 L Narrow Mouth Amber Glass Bottle 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15953077 Supplier Avantor J.T.Baker Supplier No. 563002
 Restricted Item
Estimated Shipment: 26-08-2026
Add to basket
This item is not returnable. View return policy

High quality chemicals for laboratory and specialized industrial use

TRUSTED_SUSTAINABILITY

Specifications

CAS 7761-88-8
Quantity 1 L
Molecular Formula AgNO3
MDL Number MFCD00003414
Synonym silver nitrate, silvernitrate, lunar caustic, silbernitrat, argenti nitras, nitrate d'argent, nitric acid silver i salt, silver mononitrate, silver i nitrate, argerol
InChI Key SQGYOTSLMSWVJD-UHFFFAOYSA-N
SMILES [Ag+].[O-][N+]([O-])=O
IUPAC Name silver(1+) nitrate
Molecular Weight (g/mol) 169.87
PubChem CID 24470
ChEBI CHEBI:32130
Formula Weight 169.87
Grade BAKER ANALYZED™ Reagent
Packaging Narrow Mouth Amber Glass Bottle
Specific Gravity 1L = 1.00kg
Chemical Name or Material Silver Nitrate, 0.1 N Volumetric Solution
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.