Learn More
Buerkle™ Single Mark Plastic Pipets
Pipettes made of plastic are particularly suited for training purposes, in industrial labs, during field research and applications where pipettes made of glass cannot be used, e.g. in flourine and silicate chemistry
Brand:  Buerkle™ 7553-0025
26.20 EUR valid until 2024-12-31
Use promo code "23267" to get your promotional price.
Additional Details : Weight : 0.00950kg
 View more versions of this product
Product Code. 10000092
  • € 29.35 / Each
Estimated Shipment: 21-05-2024
to see stock.
Add to basket
Description
Description
  • PP, transparent
  • Blue, permanently coloured graduation and labelling
Specifications
Specifications
Pipette
For Training Purposes, in Industrial Labs, During Field Research and Applications Where Pipettes Made of Glass Cannot be Used, e.g. in Fluorine and Silicate Chemistry
1
25 mL
Polypropylene
Yes
Blue
One-mark Pipet
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
Product Identifier
  • Vollpipette
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 1
Hazard Statement
  • H301-Toxic if swallowed.
  • H317-May cause an allergic skin reaction.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H317-May cause an allergic skin reaction.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Precautionary Statement