Learn More
Buerkle™ Single Mark Plastic Pipets
Pipettes made of plastic are particularly suited for training purposes, in industrial labs, during field research and applications where pipettes made of glass cannot be used, e.g. in flourine and silicate chemistry
Brand: Buerkle™ 7553-0010
25.16 EUR valid until 2024-12-31
Use promo code "23267" to get your promotional price.
Additional Details : Weight : 0.00850kg
Description
- PP, transparent
- Blue, permanently coloured graduation and labelling
Specifications
Pipette | |
For Training Purposes, in Industrial Labs, During Field Research and Applications Where Pipettes Made of Glass Cannot be Used, e.g. in Fluorine and Silicate Chemistry | |
1 | |
10 mL |
Polypropylene | |
Yes | |
Transparent | |
One-mark Pipet |