Learn More
Honeywell™ SKELETON NIT 3 Gloves
Click to view available options
Size:
8
9
10
Unit Size:
1 pair
Description
- Knitted glove in Spectra™
- The knitting technique provides the glove with excellent fit and high dexterity
- The Spectra™ yarn offers very good resistance to cut
- Black nitrile foam coating on palm
- Nitrile coating provides good protection against oil and grease
- The foam coating makes the glove more flexible than standard nitrile
- Rubber pad on top of the hand and fingers
- Helps to protect metacarpian and fingers from shocks
Specifications
Specifications
| Certifications/Compliance | EN 420-03 EEC Basic standards, EN 388-03 Mechanical hazards |
| Description | 13 gauge |
| Color | Blue, White |
| Material | Nitrile |
| Size | 8 |
| Length (Metric) | 25 cm |