Learn More
SLA/LP Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 11551-1-AP-20UL
 View more versions of this product
Product Code. 16818713
  • € 145.00 / 20µL
Description
SLAP also known as Src-like-adapter protein 1 is a 276 amino acid containing adapter protein, functioning as a negative regulator of T-cell receptor (TCR) signaling. SLAP belonging to the SRC family of cytoplasmic tyrosine kinases contains a SH2 domain and a SH3 domain. This cytoplasmic protein colocalizes with endosomes and is known to inhibit T-cell antigen-receptor induced activation of nuclear factor of activated T-cells. It acts as a negative regulator of positive selection and mitosis of T-cells. SLAP may act by linking signaling proteins such as ZAP70 with CBL, leading to a CBL dependent degradation of signaling proteins. Expression of SLAP is weak in heart, adult brain, placenta, liver, skeletal muscle, kidney and pancreas but is high in lung and fetal brain.
Specifications
Specifications
SLA/LP
Polyclonal
Unconjugated
SEPSECS
Liver pancreas antigen, LP, Sec synthase, Selenocysteine synthase, Sep tRNA:Sec tRNA synthase, SEPSECS, SLA, SLA p35, SLA/LP, SLA/LP autoantigen, Soluble liver antigen, TRNP48
Rabbit
Antigen Affinity Chromatography
RUO
211006, 51091, 679383
-20°C
Liquid
Western Blot, Immunoprecipitation
0.15 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q6P6M7, Q9HD40
SEPSECS
SLA/LP Fusion Protein Ag2097
20 μL
Primary
Rat, Human, Mouse