missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLC12A3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 1 publication
Brand: Novus Biologicals NBP1-44270-0.025ml
This item is not returnable.
View return policy
Description
SLC12A3 Polyclonal specifically detects SLC12A3 in Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Electron Microscopy.
Specifications
| SLC12A3 | |
| Polyclonal | |
| Unconjugated | |
| PBS and 50% Glycerol with 0.09% Sodium Azide | |
| FLJ96318, Na-Cl symporter, NCCT, solute carrier family 12 (sodium/chloride transporters), member 3, solute carrier family 12 member 3, thiazide-sensitive Na-Cl cotransporter, Thiazide-sensitive sodium-chloride cotransporter, TSCNaCl electroneutral thiazide-sensitive cotransporter | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 6559 | |
| Mouse | |
| Purified |
| Western Blot | |
| 1 mg/mL | |
| Western Blot 1:1000, Immunohistochemistry 1:200, Immunocytochemistry/Immunofluorescence 1:10-1:2000, Immunohistochemistry-Paraffin 1:200, Electron Microscopy 1:10-1:500 | |
| NP_062218 | |
| SLC12A3 | |
| Poduced against a synthetic peptide mapping to a segment of rat NCC (amino acids 74-95), N-terminal | |
| 0.025 mL | |
| Primary | |
| Detects approx 160kDa. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction