missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLC12A3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 1 publication
€ 158.00
Specifications
| Antigen | SLC12A3 |
|---|---|
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:1000, Immunohistochemistry 1:200, Immunocytochemistry/Immunofluorescence 1:10-1:2000, Immunohistochemistry-Paraffin 1:200, Electron Microscopy 1:10-1:500 |
| Applications | Western Blot |
| Classification | Polyclonal |
Description
SLC12A3 Polyclonal specifically detects SLC12A3 in Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Electron Microscopy.Specifications
| SLC12A3 | |
| Western Blot 1:1000, Immunohistochemistry 1:200, Immunocytochemistry/Immunofluorescence 1:10-1:2000, Immunohistochemistry-Paraffin 1:200, Electron Microscopy 1:10-1:500 | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS and 50% Glycerol with 0.09% Sodium Azide | |
| FLJ96318, Na-Cl symporter, NCCT, solute carrier family 12 (sodium/chloride transporters), member 3, solute carrier family 12 member 3, thiazide-sensitive Na-Cl cotransporter, Thiazide-sensitive sodium-chloride cotransporter, TSCNaCl electroneutral thiazide-sensitive cotransporter | |
| SLC12A3 | |
| IgG | |
| Affinity purified | |
| Detects approx 160kDa. |
| 1 mg/mL | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Mouse | |
| NP_062218 | |
| 6559 | |
| Poduced against a synthetic peptide mapping to a segment of rat NCC (amino acids 74-95), N-terminal | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title