Learn More
SLC14A2 Rabbit, anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 21932-1-AP-150UL
 View more versions of this product
Product Code. 18005165
  • € 410.00 / 150µL
Description
The SLC14A2 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human SLC14A2. This antibody recognizes human, mouse antigen. The SLC14A2 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
Specifications
SLC14A2
350 μg/mL
Rabbit
Primary
Human, Mouse
150 kDa
IgG
Western Blot, ELISA
hUT A6, HUT2, SLC14A2, Urea transporter 2, Urea transporter, kidney, UT A2, UT2, UTA, UTR
150 μL
8170
Store at -20°C. Stable for one year after shipment.