Learn More
SLC22A2/OCT2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 2 publications
Brand:  Novus Biologicals NBP1-89417
 View more versions of this product
Product Code. 18711214
  • € 539.00 / 0.10mL
Description
SLC22A2/OCT2 Polyclonal specifically detects SLC22A2/OCT2 in Human, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Single Cell Western.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Western Blot, Immunohistochemistry
Western Blot Reactivity reported in scientific literature (PMID: 26045896)., Immunohistochemistry 1:500 - 1:1000, Immunohistochemistry-Paraffin 1:500 - 1:1000, Single Cell Western Single Cell Western reported by an internal validation at a 1:20 dilution
Affinity Purified
RUO
IgG
Polyclonal
63 kDa
0.1 mL
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.