Learn More
SLC22A2/OCT2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 2 publications
Brand: Novus Biologicals NBP1-89417
This item is not returnable.
View return policy
Description
SLC22A2/OCT2 Polyclonal specifically detects SLC22A2/OCT2 in Human, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Single Cell Western.
Specifications
| Western Blot, Immunohistochemistry | |
| Western Blot Reactivity reported in scientific literature (PMID: 26045896)., Immunohistochemistry 1:500 - 1:1000, Immunohistochemistry-Paraffin 1:500 - 1:1000, Single Cell Western Single Cell Western reported by an internal validation at a 1:20 dilution | |
| Affinity Purified | |
| RUO | |
| IgG |
| Polyclonal | |
| 63 kDa | |
| 0.1 mL | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |