missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLC22A7 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 26796-1-AP-20UL
This item is not returnable.
View return policy
Description
SLC22A7 is involved in the sodium-independent transport and excretion of organic anions, some of which are potentially toxic. The protein is an integral membrane protein and appears to be localized to the basolateral membrane of the kidney.The protein encoded by this gene is involved in the sodium-independent transport and excretion of organic anions, some of which are potentially toxic. The encoded protein is an integral membrane protein and appears to be localized to the basolateral membrane of the kidney. Alternatively spliced transcript variants encoding different isoforms have been described.The protein encoded by this gene is involved in the sodium-independent transport and excretion of organic anions, some of which are potentially toxic. The encoded protein is an integral membrane protein and appears to be localized to the basolateral membrane of the kidney. Alternatively spliced transcript variants encoding different isoforms have been described.Specifications
| SLC22A7 | |
| Polyclonal | |
| Unconjugated | |
| SLC22A7 | |
| hOAT2, NLT, Novel liver transporter, OAT2, Organic anion transporter 2, SLC22A7 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 108114, 10864, 89776 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunoprecipitation | |
| 0.37 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q5RLM2, Q91WU2, Q9Y694 | |
| Slc22a7 | |
| SLC22A7 Fusion Protein Ag25217 | |
| 20 μL | |
| Primary | |
| Mouse, Human, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction