missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLC22A7 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
€ 145.00 - € 410.00
Specifications
| Antigen | SLC22A7 |
|---|---|
| Concentration | 0.37 mg/mL |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunoprecipitation |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16885155
|
Proteintech
26796-1-AP-20UL |
20 μL |
€ 145.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16875155
|
Proteintech
26796-1-AP-150UL |
150 μL |
€ 410.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
SLC22A7 is involved in the sodium-independent transport and excretion of organic anions, some of which are potentially toxic. The protein is an integral membrane protein and appears to be localized to the basolateral membrane of the kidney.The protein encoded by this gene is involved in the sodium-independent transport and excretion of organic anions, some of which are potentially toxic. The encoded protein is an integral membrane protein and appears to be localized to the basolateral membrane of the kidney. Alternatively spliced transcript variants encoding different isoforms have been described.The protein encoded by this gene is involved in the sodium-independent transport and excretion of organic anions, some of which are potentially toxic. The encoded protein is an integral membrane protein and appears to be localized to the basolateral membrane of the kidney. Alternatively spliced transcript variants encoding different isoforms have been described.Specifications
| SLC22A7 | |
| Immunohistochemistry (Paraffin), Western Blot, Immunoprecipitation | |
| Unconjugated | |
| Rabbit | |
| Mouse, Human, Rat | |
| Q5RLM2, Q91WU2, Q9Y694 | |
| 108114, 10864, 89776 | |
| SLC22A7 Fusion Protein Ag25217 | |
| Primary | |
| -20°C | |
| SLC22A7 |
| 0.37 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| hOAT2, NLT, Novel liver transporter, OAT2, Organic anion transporter 2, SLC22A7 | |
| Slc22a7 | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title