missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLC36A4 Antibody (PAT-4/9/H10) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
€ 214.00 - € 382.00
Specifications
| Antigen | SLC36A4 |
|---|---|
| Clone | PAT-4/9/H10 |
| Concentration | 1.1 mg/mL |
| Dilution | Western Blot 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500 |
| Applications | Western Blot, Immunohistochemistry |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18617898
|
Novus Biologicals
NBP2-50237-0.025ml |
0.025 mL |
€ 214.00
0.03mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18602709
|
Novus Biologicals
NBP2-50237 |
0.1 mL |
€ 382.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
SLC36A4 Monoclonal antibody specifically detects SLC36A4 in Human samples. It is validated for Western Blot, ImmunohistochemistrySpecifications
| SLC36A4 | |
| 1.1 mg/mL | |
| Western Blot, Immunohistochemistry | |
| Unconjugated | |
| Mouse | |
| Cellular Signaling, metabolism | |
| PBS | |
| 120103 | |
| IgG2a κ | |
| Protein A purified |
| PAT-4/9/H10 | |
| Western Blot 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| proton/amino acid transporter 4, solute carrier family 36 (proton/amino acid symporter), member 4 | |
| Antigenic amino acid sequence within the N-terminus of SLC36A4 (REELDMDVMRPLINE-C). | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title