Learn More
SLC37A4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 20612-1-AP-150UL
This item is not returnable.
View return policy
Description
SLC37A4 transports glucose-6-phosphate from the cytoplasm to the lumen of the endoplasmic reticulum. It forms with glucose-6-phosphatase the complex responsible for glucose production through glycogenolysis and gluconeogenesis. Hence, it plays a central role in homeostatic regulation of blood glucose levels.Specifications
| SLC37A4 | |
| Polyclonal | |
| Unconjugated | |
| SLC37A4 | |
| G6PT, G6PT1, G6PT2, G6PT3, GSD1b, GSD1c, GSD1d, PRO0685, SLC37A4, TRG 19, TRG19 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 14385, 2542, 29573 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.18 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O43826 | |
| SLC37A4 | |
| SLC37A4 Fusion Protein Ag14660 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |