missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLC6A3/DAT1 Antibody (SR1877), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-21721-50ul
This item is not returnable.
View return policy
Description
SLC6A3/DAT1 Monoclonal antibody specifically detects SLC6A3/DAT1 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunoprecipitation
Specifications
| SLC6A3/DAT1 | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Flow Cytometry, Immunoprecipitation | |
| SR1877 | |
| Western Blot 1:1000-1:5000, Flow Cytometry 1:50, Immunoprecipitation 1:50 | |
| DA transporter, DATDAT1Solute carrier family 6 member 3, sodium-dependent dopamine transporter, solute carrier family 6 (neurotransmitter transporter, dopamine), member 3 | |
| A synthesized peptide derived from human SLC6A3/DAT1 (Uniprot #: Q01959) | |
| 50 μg | |
| Neuroscience | |
| 6531 | |
| Store at -20° C. Avoid freeze/thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction