missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLC6A3/DAT1 Antibody (SR1877), Novus Biologicals™
Rabbit Monoclonal Antibody
€ 338.00 - € 449.00
Specifications
| Antigen | SLC6A3/DAT1 |
|---|---|
| Clone | SR1877 |
| Dilution | Western Blot 1:1000-1:5000, Flow Cytometry 1:50, Immunoprecipitation 1:50 |
| Applications | Western Blot, Flow Cytometry, Immunoprecipitation |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18671975
|
Novus Biologicals
NBP3-21721-100ul |
100 μg |
€ 449.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18643744
|
Novus Biologicals
NBP3-21721-50ul |
50 μg |
€ 338.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
SLC6A3/DAT1 Monoclonal antibody specifically detects SLC6A3/DAT1 in Human samples. It is validated for Western Blot, Flow Cytometry, ImmunoprecipitationSpecifications
| SLC6A3/DAT1 | |
| Western Blot 1:1000-1:5000, Flow Cytometry 1:50, Immunoprecipitation 1:50 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| DA transporter, DATDAT1Solute carrier family 6 member 3, sodium-dependent dopamine transporter, solute carrier family 6 (neurotransmitter transporter, dopamine), member 3 | |
| A synthesized peptide derived from human SLC6A3/DAT1 (Uniprot #: Q01959) | |
| Primary | |
| Store at -20° C. Avoid freeze/thaw cycles. |
| SR1877 | |
| Western Blot, Flow Cytometry, Immunoprecipitation | |
| Unconjugated | |
| Rabbit | |
| Neuroscience | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| 6531 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title