Learn More
Edmund Bühler GmbH™ Sliding Plate with Combifix VKS
Affix to a multistory platform. Edmund Bühler GmbH™ Sliding Plate with Combifix VKS fastens horizontal separating funnels of different sizes to platform.
Brand:  Edmund Bühler GmbH™ 0051 495
Additional Details : Weight : 0.80000kg
 View more versions of this product
Product Code. 11824321
  • € 955.00 / Each
Estimated Shipment: 03-07-2024
to see stock.
Add to basket
Description
Description
  • Fastens horizontal separating funnels of different sizes to platform
  • For use with Combifix VKS or SM Universal Shakers
Specifications
Specifications
Sliding Plate
Sliding plates with Combifix VKS C
2 Clamping Strips H, 2 Clamping Strips V, 1 Spring Strip, base plate and basic rack
760 x 600mm
Combifix VKS
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers