Learn More
Edmund Bühler GmbH™ Sliding Plate with Combifix VKS
Affix to a multistory platform. Edmund Bühler GmbH™ Sliding Plate with Combifix VKS fastens horizontal separating funnels of different sizes to platform.
€ 860.00 - € 955.00
Product Code | Brand | Includes | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Includes | Price | Quantity & Availability | |||||
11804271
|
Edmund Bühler GmbH™
0051 493 |
9 Clamping Strips H, base plate and basic rack |
€ 860.00
Each |
Estimated Shipment: 03-07-2024
Log in to see stock. |
|||||
11824321
|
Edmund Bühler GmbH™
0051 495 |
2 Clamping Strips H, 2 Clamping Strips V, 1 Spring Strip, base plate and basic rack |
€ 955.00
Each |
Estimated Shipment: 26-06-2024
Log in to see stock. |
|||||
Description
- Fastens horizontal separating funnels of different sizes to platform
- For use with Combifix VKS or SM Universal Shakers
Specifications
Sliding Plate | |
Combifix VKS |