Learn More
Edmund Bühler GmbH™ Sliding Plate with Combifix VKS
Affix to a multistory platform. Edmund Bühler GmbH™ Sliding Plate with Combifix VKS fastens horizontal separating funnels of different sizes to platform.
€ 860.00 - € 955.00
Specifications
Product Type Sliding Plate
For Use With (Equipment) Combifix VKS
View More Specs
Products 2
Product Code Brand Includes Price Quantity & Availability  
Product Code Brand Includes Price Quantity & Availability  
11804271
View Documents Promotion Details
Edmund Bühler GmbH™
0051 493
9 Clamping Strips H, base plate and basic rack
€ 860.00
Each
View alternative products
Save Up to 
»
Estimated Shipment: 03-07-2024
to see stock.
Add to basket
11824321
View Documents Promotion Details
Edmund Bühler GmbH™
0051 495
2 Clamping Strips H, 2 Clamping Strips V, 1 Spring Strip, base plate and basic rack
€ 955.00
Each
View alternative products
Save Up to 
»
Estimated Shipment: 26-06-2024
to see stock.
Add to basket
Description
Description
  • Fastens horizontal separating funnels of different sizes to platform
  • For use with Combifix VKS or SM Universal Shakers
Specifications
Specifications
Sliding Plate
Combifix VKS