missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SLIT1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5119420
This item is not returnable.
View return policy
Description
SLIT1 Polyclonal Antibody for Western Blot, IHC (P)
SLIT1 is thought to act as molecular guidance cue in cellular migration, and function appears to be mediated by interaction with roundabout homolog receptors.
Specifications
| SLIT1 | |
| Polyclonal | |
| Unconjugated | |
| SLIT1 | |
| KIAA0813; Megf4; mKIAA0813; Multiple EGF-like domains protein 4; multiple epidermal growth factor-like domains 4; multiple epidermal growth factor-like domains protein 4; SLIL1; slit guidance ligand 1; slit homolog 1; slit homolog 1 (Drosophila); slit homolog 1 protein; SLIT1; Slit-1; SLIT3 | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 20562, 65047 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Western Blot | |
| 0.5 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| O88279, Q80TR4 | |
| SLIT1 | |
| Recombinant protein Slit Homolog 1. The antigen corresponds to amino acid range 308-512 of the target protein. | |
| 200 μL | |
| Primary | |
| Mouse, Rat | |
| Antibody | |
| IgG |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur