missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SLIT1 Polyclonal Antibody

Rabbit Polyclonal Antibody
€ 212.00 - € 514.00
Specifications
| Antigen | SLIT1 |
|---|---|
| Concentration | 0.5 mg/mL |
| Content And Storage | Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Western Blot |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17750824
![]() |
Invitrogen™
PA5119420 |
200 μL |
€ 514.00
200µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
17772994
|
Invitrogen™
PA5121599 |
20 μL |
€ 212.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
SLIT1 Polyclonal Antibody for Western Blot, IHC (P)
SLIT1 is thought to act as molecular guidance cue in cellular migration, and function appears to be mediated by interaction with roundabout homolog receptors.Specifications
| SLIT1 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Polyclonal | |
| Liquid | |
| IgG | |
| Mouse, Rat | |
| SLIT1 | |
| KIAA0813; Megf4; mKIAA0813; Multiple EGF-like domains protein 4; multiple epidermal growth factor-like domains 4; multiple epidermal growth factor-like domains protein 4; SLIL1; slit guidance ligand 1; slit homolog 1; slit homolog 1 (Drosophila); slit homolog 1 protein; SLIT1; Slit-1; SLIT3 | |
| SLIT1 | |
| Primary | |
| Antigen affinity chromatography, Protein A |
| 0.5 mg/mL | |
| Immunohistochemistry (Paraffin), Western Blot, Western Blot | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| O88279, Q80TR4 | |
| 20562, 65047 | |
| Recombinant protein Slit Homolog 1. The antigen corresponds to amino acid range 308-512 of the target protein. | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title