missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLP-2 Antibody - BSA Free, Novus Biologicals™
Chicken Polyclonal Antibody
Brand: Novus Biologicals NB100-75482-0.025mg
This item is not returnable.
View return policy
Description
SLP-2 Polyclonal specifically detects SLP-2 in Human, Mouse, Rat samples. It is validated for Western Blot.
Specifications
| SLP-2 | |
| Polyclonal | |
| Unconjugated | |
| Q53G29 | |
| STOML2 | |
| The immunogen for this antibody was a sequence from 171a.a. to 356a.a. from STOML2(a 356a.a. long protein) | |
| 0.025 mg | |
| Core ESC Like Genes, Stem Cell Markers | |
| 30968 | |
| Human |
| Western Blot | |
| 1 mg/mL | |
| PBS with No Preservative | |
| EPB72-like protein 2, HSPC108, SLP2, SLP-2stomatin-like protein 2, stomatin (EPB72)-like 2, stomatin-like 2 | |
| Chicken | |
| Affinity Purified | |
| RUO | |
| Primary | |
| STOML2 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction