missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLP-2 Antibody - BSA Free, Novus Biologicals™
Chicken Polyclonal Antibody
€ 370.00
Specifications
| Antigen | SLP-2 |
|---|---|
| Concentration | 1 mg/mL |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
SLP-2 Polyclonal specifically detects SLP-2 in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
| SLP-2 | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| Human | |
| Q53G29 | |
| 30968 | |
| The immunogen for this antibody was a sequence from 171a.a. to 356a.a. from STOML2(a 356a.a. long protein) | |
| Affinity Purified | |
| STOML2 |
| 1 mg/mL | |
| Polyclonal | |
| Chicken | |
| Core ESC Like Genes, Stem Cell Markers | |
| PBS with No Preservative | |
| EPB72-like protein 2, HSPC108, SLP2, SLP-2stomatin-like protein 2, stomatin (EPB72)-like 2, stomatin-like 2 | |
| STOML2 | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title