missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SM047 Antibody (SM047), Novus Biologicals™
Mouse Monoclonal Antibody has been used in 1 publication
Brand: Novus Biologicals NBP1-97921SS
This item is not returnable.
View return policy
Description
SM047 Monoclonal specifically detects SM047 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin.
Specifications
| SM047 | |
| Monoclonal | |
| Unconjugated | |
| Ascites with 0.1% Sodium Azide | |
| Mouse | |
| Unpurified | |
| RUO | |
| Primary | |
| Human | |
| Ascites |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| SM047 | |
| Western Blot 1:1000-1:5000, Immunohistochemistry 1:100-1:1000, Immunocytochemistry/Immunofluorescence 1:100-1:1000, Immunoprecipitation 1:10-1:100, Immunohistochemistry-Paraffin 1:100-1:1000 | |
| anti-SMO47 | |
| Cell membranes from human ovarian serous cystadenocarcinmoma and cells from human ovarian cancer cell line UWOV1. | |
| 0.025 mL | |
| Cancer, Ovarian Carcinoma Cell Markers | |
| Glycocalyx of human ovarian carcinoma cells. | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. | |
| IgM |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction