missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SM047 Antibody (SM047), Novus Biologicals™
Mouse Monoclonal Antibody has been used in 1 publication
€ 199.00 - € 450.00
Specifications
| Antigen | SM047 |
|---|---|
| Clone | SM047 |
| Dilution | Western Blot 1:1000-1:5000, Immunohistochemistry 1:100-1:1000, Immunocytochemistry/Immunofluorescence 1:100-1:1000, Immunoprecipitation 1:10-1:100, Immunohistochemistry-Paraffin 1:100-1:1000 |
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18049096
|
Novus Biologicals
NBP1-97921SS |
0.025 mL |
€ 199.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18094927
|
Novus Biologicals
NBP1-97921 |
0.1 mL |
€ 450.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
SM047 Monoclonal specifically detects SM047 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin.Specifications
| SM047 | |
| Western Blot 1:1000-1:5000, Immunohistochemistry 1:100-1:1000, Immunocytochemistry/Immunofluorescence 1:100-1:1000, Immunoprecipitation 1:10-1:100, Immunohistochemistry-Paraffin 1:100-1:1000 | |
| Monoclonal | |
| Ascites | |
| RUO | |
| Human | |
| anti-SMO47 | |
| IgM | |
| Unpurified | |
| Glycocalyx of human ovarian carcinoma cells. |
| SM047 | |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Cancer, Ovarian Carcinoma Cell Markers | |
| Ascites with 0.1% Sodium Azide | |
| Cell membranes from human ovarian serous cystadenocarcinmoma and cells from human ovarian cancer cell line UWOV1. | |
| Primary | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title