Learn More
SMAD7 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
€ 145.00 - € 410.00
Specifications
| Antigen | SMAD7 |
|---|---|
| Concentration | 540 μg/mL |
| Applications | Western Blot, Immunoprecipitation, Immunohistochemistry, Immunofluorescence, ELISA |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16804405
|
Proteintech
25840-1-AP-20UL |
20 μL |
€ 145.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16894395
|
Proteintech
25840-1-AP-150UL |
150 μL |
€ 410.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The SMAD7 antibody from Proteintech is a rabbit polyclonal antibody generated with fusion protein of human SMAD7. This antibody recognizes human, mouse, rat antigen. The SMAD7 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
| SMAD7 | |
| Western Blot, Immunoprecipitation, Immunohistochemistry, Immunofluorescence, ELISA | |
| Unconjugated | |
| Human, Rat, Mouse | |
| IgG |
| 540 μg/mL | |
| Polyclonal | |
| Rabbit | |
| 4092 | |
| Primary |