Learn More
SNX11 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand:  Novus Biologicals NBP1-85825
 View more versions of this product
Product Code. 18725543
  • € 589.00 / 0.10mL
Description
SNX11 Polyclonal specifically detects SNX11 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Polyclonal
RUO
IgG
This antibody was developed against Recombinant Protein corresponding to amino acids:LFLQSQLSVPEIEACVQGRSTMTVSDAILRYAMSNCGWAQEERQSSSHLAKGDQPKSCCFLPRSGRRSSP
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.