missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SNX9 Monoclonal Antibody (OTI2A11), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA501323S
This item is not returnable.
View return policy
Description
SNX9 Monoclonal antibody specifically detects SNX9 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Western BlotSpecifications
| SNX9 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q9Y5X1 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 51429 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG2a |
| Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
| OTI2A11 | |
| Unconjugated | |
| Snx9 | |
| 2700073N08Rik; MST155; MSTP155; OTTHUMP00000040083; Protein SDP1; RP11-266C7.10; SDP1; SH3 and PX domain-containing protein 1; SH3 and PX domain-containing protein 3A; SH3PX1; SH3PXD3A; Snx9; sorting nexin 9; sorting nexin-9; Wiskott-Aldrich syndrome protein (WASP) interactor protein; WISP | |
| Full length human recombinant protein of human SNX9 produced in HEK293T cell. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction