missing translation for 'onlineSavingsMsg'
Learn More

Sodium Arsenite, Reagent Grade, Ricca Chemical

Product Code. 31750907
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
31750907 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Product Code. 31750907 Supplier Ricca Chemical Company Supplier No. RDCS0280100B1
 Restricted Item
Estimated Shipment: 14-10-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

Specifications

CAS 7784-46-5
CAS Min % 98.0
CAS Max % 100.0
Physical Form Fine Crystals or Crystalline Powder
Quantity 100 g
Molecular Formula AsNaO2
Synonym sodium arsenite, sodium metaarsenite, sodium dioxoarsenate, prodalumnol, sodanit, penite, kill-all, prodalumnol double, rat death liquid, chem pels c
InChI Key PTLRDCMBXHILCL-UHFFFAOYSA-M
SMILES [O-][As]=O.[Na+]
IUPAC Name sodium;oxoarsinite
Molecular Weight (g/mol) 129.909
PubChem CID 443495
ChEBI CHEBI:29678
Percent Purity ≥ 98%
Grade Reagent
Packaging White Poly Wide Mouth Bottle
Chemical Name or Material Sodium Arsenite
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.