Learn More
Sodium Borohydride, TRC
Product Code. 30006794
Change view
Click to view available options
Quantity:
5 g
100 g
Unit Size:
100g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30006794 100 g 100g
30005947 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30006794 Supplier Toronto Research Chemicals Supplier No. TRCS614900100G
This item requires a price update prior to being ordered.
Request a Price for this item here.
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 16940-66-2
Molecular Formula BH4Na
Molecular Weight (g/mol) 37.83
Synonym Tetrahydroborate(1-) Sodium, Borol, Colorstrip 2000, Hidkitex DF, Hydrifin, Sodium Borohydrate, Sodium Borohydride, Sodium Borohydride, NaBH4, Sodium Hydroborate, Sodium Tetrahydridoborate, Sodium tetrahydroborate, Sodium Tetrahydroborate(1-), VenMet
IUPAC Name sodium;boranuide
SMILES [BH4-].[Na+]
Specifications
Quantity 100 g
InChI Formula InChI=1S/BH4.Na/h1H4;/q-1;+1
Formula Weight 38.0304
Chemical Name or Material Sodium Borohydride