missing translation for 'onlineSavingsMsg'
Learn More

Sodium chloride, TraceSELECT™, Honeywell Fluka™

Product Code. p-7072199 Shop All Honeywell Fluka Products
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
This item is not returnable. View return policy

Product Code. 15625350

Brand: Honeywell-Fluka 3897925G

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 7647-14-5
Molecular Formula ClNa
Molecular Weight (g/mol) 58.44
MDL Number MFCD00003477
InChI Key FAPWRFPIFSIZLT-UHFFFAOYSA-M
Synonym sodium chloride, salt, table salt, halite, saline, rock salt, common salt, sodium chloride nacl, dendritis, purex
PubChem CID 5234
ChEBI CHEBI:26710
IUPAC Name sodium chloride
SMILES [Na+].[Cl-]

Specifications

Quantity 25 g
Linear Formula NaCl
UN Number NONH for all modes of transport
Beilstein 3534976
Formula Weight 58.44g/mol
Percent Purity ≥99.99% (metals basis)
Grade TraceSELECT™
Vapor Pressure 1.33 (1.00 mmHg) at 865°C (1589°F) hPa
Chemical Name or Material Sodium chloride
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.