missing translation for 'onlineSavingsMsg'
Learn More

Sodium hexafluorophosphate, 98%

Product Code. 11459936
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 11459936

Brand: Thermo Scientific Alfa Aesar B20898.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 21324-39-0 | F6NaP | 167.95 g/mol

Sodium hexafluorophosphate is used as an ion pairing agent for the determination of cobalt(II) in water samples. It is used as an important raw material in the preparation of 2-chloro-1,3-bis(dimethylamino)trimethinium hexafluorophosphate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium hexafluorophosphate is used as an ion pairing agent for the determination of cobalt(II) in water samples. It is used as an important raw material in the preparation of 2-chloro-1,3-bis(dimethylamino)trimethinium hexafluorophosphate.

Solubility
Soluble in water.

Notes
Hygroscopic. Incompatible with strong acids, bases and oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 21324-39-0
Molecular Formula F6NaP
Molecular Weight (g/mol) 167.95
MDL Number MFCD00011122
InChI Key KMADQUOCJBLXRP-UHFFFAOYSA-N
Synonym sodium hexafluorophosphate, sodium hexa-fluoro phosphate, sodium hexafluorophosphate v, napf6, sodium hexaflurophosphate, acmc-1cao2, sodium hexafluoro-, e5-phosphanuide, sodium hexafluoro-$l^ 5-phosphanuide, phosphate 1-, hexafluoro-, sodium 1:1
PubChem CID 5147921
SMILES [Na+].F[P-](F)(F)(F)(F)F

Specifications

Melting Point >200°C
Quantity 25 g
UN Number UN3260
Solubility Information Soluble in water.
IUPAC Name sodium hexafluoro-λ⁵-phosphanuide
Formula Weight 167.95
Percent Purity 98%
Odor Odorless
Sensitivity Hygroscopic
Density 2.37
Chemical Name or Material Sodium hexafluorophosphate
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Sodium hexafluorophosphate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H332-Harmful if inhaled.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : Sodium hexafluorophosphate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.