Learn More
Sodium Peroxide, TRC
Product Code. 30428356
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30428356 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30428356 Supplier Toronto Research Chemicals Supplier No. TRCS665800100G
This item requires a price update prior to being ordered.
Request a Price for this item here.
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Sodium peroxide
CAS 1313-60-6
Quantity 100 g
Molecular Formula Na O2
Synonym Disodium Dioxide, Disodium Peroxide, Sodium Oxide (Na2O2)
InChI Formula InChI=1S/Na.O2/c;1-2/q+1;-2
SMILES [Na+].[O-][O-]
IUPAC Name sodium;peroxide
Molecular Weight (g/mol) 54.9891
Formula Weight 54.9801