missing translation for 'onlineSavingsMsg'
Learn More

Sodium tetrachloropalladate(II), approx. 36% Pd

Product Code. p-42437 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 10241832

Brand: Thermo Scientific Acros 195130010

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 13820-53-6 | Cl4Na2Pd | 294.20 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13820-53-6
Molecular Formula Cl4Na2Pd
Molecular Weight (g/mol) 294.20
MDL Number MFCD00003487
InChI Key ABKQFSYGIHQQLS-UHFFFAOYSA-J
Synonym sodium tetrachloropalladate ii, disodium tetrachloropalladate, palladium ii sodium chloride, sodiumpalladiumchloride, disodiumpalladiumtetrachloride, sodiumtetrachloropalladate ii, palladiumsodiumchloridetrihydrate, disodiumtetrachloropalladate 2-, tetrachloro-palladate 2-disodium, disodium;tetrachloropalladium 2-
PubChem CID 11000870
SMILES [Na+].[Na+].Cl[Pd++](Cl)(Cl)Cl

Specifications

Color Brown to Orange or Brown-Green
Physical Form Powder and Granules, Crystals
Quantity 1 g
Assay Percent Range 36%
Linear Formula Na2PdCl4
Fieser 05,626; 06,45; 09,207; 16,309; 01,778; 04,365; 05,498; 06,443; 07,274; 15,13; 16,356
Solubility Information Solubility in water: soluble
IUPAC Name disodium;tetrachloropalladium(2-)
Formula Weight 294.21
Percent Purity ∼36% Pd
Packaging Glass bottle
Chemical Name or Material Sodium tetrachloropalladate(II), ca. 36.4% Pd
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Sodium tetrachloropalladate(II)
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Substances and mixtures corrosive to metals Category 1
Hazard Statement
  • H319-Causes serious eye irritation.
  • H302-Harmful if swallowed.
  • H290-May be corrosive to metals.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.