missing translation for 'onlineSavingsMsg'
Learn More

Honeywell Safety Products™ Sofraclean Gloves

Product Code. 12627543
Change view
Click to view available options
:
Size: 7
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
12627543 Size: 7
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12627543 Supplier Honeywell Safety Products™ Supplier No. 207822207

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Tightly knit cotton interlock construction gives these ambidextrous gloves properties well suited to fine work in electronics and related industries. Honeywell Safety Products™ Sofraclean Gloves provide great touch, comfort and fit.

  • Tight knit cotton interlock construction provides the following:
    • Support.
    • Touch.
    • Sweat absorption.
    • Mechanical resistance.
    • Limited dirt penetration.
  • Ribbed cotton wrist for comfort and fit.
  • Ambidextrous.
  • Ecru color enhances gloves hygiene and cleanliness properties.
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance CE CATEGORY I Minor risks, En 420-03 Basic standards, EC Category PPE 1
Classification Dexterity: 5
Cuff Material Cotton
Color Beige
Hand Ambidextrous
Material Cotton
Product Type Gloves
Size 7
For Use With (Application) Aviation and Telecom industries.
Quantity 10Pair/PK
Resistant To Cuts
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.