Learn More
SorLA Antibody (SR1433), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand:  Novus Biologicals NBP3-22079-50ul
 View more versions of this product
Product Code. 18683315
  • € 338.00 / 50µL
Description
SorLA Monoclonal antibody specifically detects SorLA in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry
TRUSTED_SUSTAINABILITY
Specifications
Specifications
SorLA
Monoclonal
Unconjugated
PBS, pH 7.4, 150mM NaCl, 50% glycerol.
Rabbit
Affinity purified
RUO
6653
Store at -20° C. Avoid freeze/thaw cycles.
IgG
Western Blot, Immunohistochemistry
SR1433
Western Blot 1:500-1:2000, Immunohistochemistry 1:100-1:500
C11orf32, chromosome 11 open reading frame 32, FLJ21930, FLJ39258, gp250, Low-density lipoprotein receptor relative with 11 ligand-binding repeats, LR11LDLR relative with 11 ligand-binding repeats, LRP9, mosaic protein LR11, SorLA, SorLA-1SORLA, sortilin-related receptor, sortilin-related receptor, L(DLR class) A repeats containing, sortilin-related receptor, L(DLR class) A repeats-containing, Sorting protein-related receptor containing LDLR class A repeats
A synthesized peptide derived from human SorLA (Uniprot #: Q92673)