missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SorLA Antibody (SR1433), Novus Biologicals™
Rabbit Monoclonal Antibody
€ 338.00 - € 449.00
Specifications
| Antigen | SorLA |
|---|---|
| Clone | SR1433 |
| Dilution | Western Blot 1:500-1:2000, Immunohistochemistry 1:100-1:500 |
| Applications | Western Blot, Immunohistochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18614365
|
Novus Biologicals
NBP3-22079-100ul |
100 μg |
€ 449.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18683315
|
Novus Biologicals
NBP3-22079-50ul |
50 μg |
€ 338.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
SorLA Monoclonal antibody specifically detects SorLA in Human, Mouse, Rat samples. It is validated for Western Blot, ImmunohistochemistrySpecifications
| SorLA | |
| Western Blot 1:500-1:2000, Immunohistochemistry 1:100-1:500 | |
| Monoclonal | |
| Purified | |
| RUO | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| 6653 | |
| IgG | |
| Affinity purified |
| SR1433 | |
| Western Blot, Immunohistochemistry | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| C11orf32, chromosome 11 open reading frame 32, FLJ21930, FLJ39258, gp250, Low-density lipoprotein receptor relative with 11 ligand-binding repeats, LR11LDLR relative with 11 ligand-binding repeats, LRP9, mosaic protein LR11, SorLA, SorLA-1SORLA, sortilin-related receptor, sortilin-related receptor, L(DLR class) A repeats containing, sortilin-related receptor, L(DLR class) A repeats-containing, Sorting protein-related receptor containing LDLR class A repeats | |
| A synthesized peptide derived from human SorLA (Uniprot #: Q92673) | |
| Primary | |
| Store at -20° C. Avoid freeze/thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title