missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SPACA5B Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 415.00 - € 624.00
Specifications
| Antigen | SPACA5B |
|---|---|
| Dilution | Immunohistochemistry 1:500 - 1:1000, Immunohistochemistry-Paraffin 1:500 - 1:1000 |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18629585
|
Novus Biologicals
NBP2-46703 |
0.1 mL |
€ 624.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18666565
|
Novus Biologicals
NBP2-46703-25ul |
25 μL |
€ 415.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
SPACA5B Polyclonal antibody specifically detects SPACA5B in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
| SPACA5B | |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Human | |
| Lysozyme-Like Protein 5, LYZL5, LYZL5B, SLLP-X, SPACA5 SPACA5B, SPACA5A, Sperm Acrosome Associated 5B, Sperm Acrosome-Associated Protein 5, Sperm-Specific Lysozyme-Like Protein X | |
| This antibody was developed against Recombinant Protein corresponding to amino acids: AGLNGYKGYGVGDWLCMAHYESGFDTAFVDHNPDGSSEYGIFQLNSAWWCDNGITPTKNLC | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Immunohistochemistry 1:500 - 1:1000, Immunohistochemistry-Paraffin 1:500 - 1:1000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS (pH 7.2), 40% Glycerol | |
| 729201 | |
| IgG | |
| Immunogen affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title