Learn More
Poulten & Graf GmbH Spare Piston for FORTUNA™ OPTIFIX™ SOLVENT Dispenser
Poulten & Graf GmbH Spare Piston for FORTUNA™ OPTIFIX™ SOLVENT Dispenser comes with a finger guard and volume setting system.
Brand:  Poulten & Graf GmbH 101 00158
Additional Details : Weight : 0.10000kg
 View more versions of this product
Product Code. 10638022
  • € 1240.00 / Each
Estimated Shipment: 29-05-2024
to see stock.
Add to basket
Description
Description
  • Made of glass
  • With finger guard and volume setting system
Specifications
Specifications
500 mL
500 mL
1
Autoclavable
Non-sterile
Piston for OPTIFIX SOLVENT Dispenser
1
Spare part for bottle top dispenser
Reusable