Learn More
Poulten & Graf GmbH Spare Piston for FORTUNA™ OPTIFIX™ SOLVENT Dispenser
Poulten & Graf GmbH Spare Piston for FORTUNA™ OPTIFIX™ SOLVENT Dispenser comes with a finger guard and volume setting system.
Brand: Poulten & Graf GmbH 101 00158
Additional Details : Weight : 0.10000kg
Description
- Made of glass
- With finger guard and volume setting system
Specifications
500 mL | |
500 mL | |
1 | |
Autoclavable | |
Non-sterile |
Piston for OPTIFIX SOLVENT Dispenser | |
1 | |
Spare part for bottle top dispenser | |
Reusable |