missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Spectrin Alpha 1 (Erythrocyte Marker) Mouse, Clone: SPTA1/1810, enQuireBio™
Mouse Monoclonal Antibody
Brand: EnquireBio 6708-MSM1-P1
Additional Details : Weight : 0.01000kg
Specifications
Spectrin Alpha 1 (Erythrocyte Marker) | |
Cytoplasmic and Cell Surface | |
SPTA1/1810 | |
Unconjugated | |
Mouse | |
240kDa | |
Protein A/G purified from Bioreactor concentrate. | |
RUO | |
6708 | |
IgG1 κ |
Immunohistochemistry, ELISA | |
Monoclonal | |
200μg/mL | |
10mM PBS with 0.05% BSA and 0.05% azide. Also available without BSA and Azide. | |
Recombinant human SPTA fragment (apprx. aa356-475) | |
Jurkat cells. Placenta or Tonsil. | |
100 μg | |
Primary | |
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. |