missing translation for 'onlineSavingsMsg'
Learn More

Spectroflux 100, Lithium tetraborate

Product Code. 11356438
Change view
Click to view available options
Quantity:
1 kg
5 x 1 kg
Unit Size:
1kg
5kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11356438 5 x 1 kg 5kg
11346438 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11356438 Supplier Thermo Scientific Alfa Aesar Supplier No. 012078.D9
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 12007-60-2

A general purpose acidic flux for fusing basic oxides, carbonates, alumino-silicates, glass, ceramics, cement steel, aluminum, concrete mixes, soils, magnesites, bauxites and rare earth oxides. Dissolves most refractories. Not suitable for highly acidic samples. Samples must be fully oxidized.Spectroflux 100, Lithium tetraborate is used in enamels. Further, it is a spectroscopic fluxing agent which is utilized for fusing basic oxides, glass, alumino-silicates, ceramics, carbonates, cement, steel, aluminum, soil, concrete mixes, bauxites, magnesites and rare earth oxides. In addition to this, it is used for dissolution of samples containing a high concentration of basic oxides and carbonate rocks and aluminosilicates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

A general purpose acidic flux for fusing basic oxides, carbonates, alumino-silicates, glass, ceramics, cement steel, aluminum, concrete mixes, soils, magnesites, bauxites and rare earth oxides. Dissolves most refractories. Not suitable for highly acidic samples. Samples must be fully oxidized.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12007-60-2

Specifications

Melting Point 930°C
Physical Form Solid
Quantity 5 x 1 kg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.