missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SPEM1 Rabbit anti-Human, Rat, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 204.00 - € 470.00
Specifications
| Antigen | SPEM1 |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Form | Purified |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18388724
|
Novus Biologicals
NBP3-04838-20UL |
20 μL |
€ 204.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18334056
|
Novus Biologicals
NBP3-04838-100UL |
100 μL |
€ 470.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
SPEM1 Polyclonal specifically detects SPEM1 in Human, Rat samples. It is validated for Western Blot.Specifications
| SPEM1 | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS with 50% glycerol, pH7.3. | |
| 374768 | |
| IgG | |
| Affinity purified |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human, Rat | |
| C17orf83, chromosome 17 open reading frame 83, FLJ40081, spermatid maturation 1, spermatid maturation protein 1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 203-309 of human SPEM1 (NP_955371.2). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title