missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SPINLW1 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-05629-20ul
This item is not returnable.
View return policy
Description
SPINLW1 Polyclonal antibody specifically detects SPINLW1 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| SPINLW1 | |
| Polyclonal | |
| Western Blot 1:500 - 1:1000 | |
| Cancer/testis antigen 71, cancer/testis antigen 72, CT71, CT72, dJ461P17.2, Epididymal protease inhibitor, eppin, EPPIN3, Protease inhibitor WAP7, serine peptidase inhibitor-like, with Kunitz and WAP domains 1 (eppin), Serine protease inhibitor-like with Kunitz and WAP domains 1, serine protease inhibitor-like, with Kunitz and WAP domains 1 (eppin), WAP four-disulfide core domain protein 7, WAP7EPPIN1, WFDC7EPPIN2 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 22-133 of human SPINLW1 (NP_065131.1). PGLTDWLFPRRCPKIREECEFQERDVCTKDRQCQDNKKCCVFSCGKKCLDLKQDVCEMPKETGPCLAYFLHWWYDKKDNTCSMFVYGGCQGNNNNFQSKANCLNTCKNKRFP | |
| 20 μg | |
| Cell Biology | |
| 57119 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction