missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SPINLW1 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 201.00 - € 463.00
Specifications
| Antigen | SPINLW1 |
|---|---|
| Dilution | Western Blot 1:500 - 1:1000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18665726
|
Novus Biologicals
NBP3-05629-100ul |
100 μg |
€ 463.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18616168
|
Novus Biologicals
NBP3-05629-20ul |
20 μg |
€ 201.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
SPINLW1 Polyclonal antibody specifically detects SPINLW1 in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
| SPINLW1 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Cell Biology | |
| PBS with 50% glycerol, pH7.3. | |
| 57119 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500 - 1:1000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| Cancer/testis antigen 71, cancer/testis antigen 72, CT71, CT72, dJ461P17.2, Epididymal protease inhibitor, eppin, EPPIN3, Protease inhibitor WAP7, serine peptidase inhibitor-like, with Kunitz and WAP domains 1 (eppin), Serine protease inhibitor-like with Kunitz and WAP domains 1, serine protease inhibitor-like, with Kunitz and WAP domains 1 (eppin), WAP four-disulfide core domain protein 7, WAP7EPPIN1, WFDC7EPPIN2 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 22-133 of human SPINLW1 (NP_065131.1). PGLTDWLFPRRCPKIREECEFQERDVCTKDRQCQDNKKCCVFSCGKKCLDLKQDVCEMPKETGPCLAYFLHWWYDKKDNTCSMFVYGGCQGNNNNFQSKANCLNTCKNKRFP | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title