missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SPRY4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5111419
This item is not returnable.
View return policy
Description
Immunogen sequence: SSDQRLLDHM APPPVADQAS PRAVRIQPKV VHCQPLDLKG PAVPPELDKH FLLCEACG Highest antigen sequence indentity to the following orthologs: Mouse - 91%, Rat - 91%.
SPRY4 is an inhibitor of the receptor-transduced mitogen-activated protein kinase (MAPK) signaling pathway. The protein is positioned upstream of RAS activation and impairs the formation of active GTP-RAS.
Specifications
| SPRY4 | |
| Polyclonal | |
| Unconjugated | |
| SPRY4 | |
| A030006O18Rik; HH17; Protein sprouty homolog 4; Sprouty 4; sprouty homolog 4; sprouty homolog 4 (Drosophila); sprouty RTK signaling antagonist 4; sprouty4; Spry4; spry-4 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 81848 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 0.10 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9C004 | |
| SPRY4 | |
| Recombinant Protein corresonding to Human SPRY4. Recombinant protein control fragment (Product #RP-105014). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction