missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SPRY4 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

€ 618.00 - € 620.00

Specifications

Antigen SPRY4
Concentration 0.10 mg/mL
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
17189333
GREENER_CHOICE
View Documents
Invitrogen™
PA5111419
100 μL
€ 620.00
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
17169613
GREENER_CHOICE
View Documents
Invitrogen™
PA5111699
100 μL
€ 618.00
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Immunogen sequence: SSDQRLLDHM APPPVADQAS PRAVRIQPKV VHCQPLDLKG PAVPPELDKH FLLCEACG Highest antigen sequence indentity to the following orthologs: Mouse - 91%, Rat - 91%.

SPRY4 is an inhibitor of the receptor-transduced mitogen-activated protein kinase (MAPK) signaling pathway. The protein is positioned upstream of RAS activation and impairs the formation of active GTP-RAS.
Specifications

Specifications

SPRY4
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Polyclonal
Liquid
IgG
Human
SPRY4
A030006O18Rik; HH17; Protein sprouty homolog 4; Sprouty 4; sprouty homolog 4; sprouty homolog 4 (Drosophila); sprouty RTK signaling antagonist 4; sprouty4; Spry4; spry-4
SPRY4
Primary
Antigen affinity chromatography
0.10 mg/mL
Western Blot, Immunocytochemistry
Unconjugated
Rabbit
RUO
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Q9C004
81848
Recombinant Protein corresonding to Human SPRY4. Recombinant protein control fragment (Product #RP-105014).
Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.