missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SQLE Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-93808-0.1ml
This item is not returnable.
View return policy
Description
SQLE Polyclonal antibody specifically detects SQLE in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| SQLE | |
| Polyclonal | |
| Western Blot 1:500 - 1:1000, Immunohistochemistry 1:50-1:100, Immunocytochemistry/ Immunofluorescence 1:50-1:100, Immunohistochemistry-Paraffin | |
| EC 1.14.99.7, ERG1, FLJ30795, squalene epoxidaseSE, squalene monooxygenase | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 300-400 of human SQLE (NP_003120.2). VSVSSHFVGFLMKNAPQFKANHAELILANPSPVLIYQISSSETRVLVDIRGEMPRNLREYMVEKIYPQIPDHLKEPFLEATDNSHLRSMPASFLPPSSVKK | |
| 0.1 mL | |
| Core ESC Like Genes, Stem Cell Markers | |
| 6713 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction