missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Src Antibody (21H5), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-26303-50ul
This item is not returnable.
View return policy
Description
Src Monoclonal antibody specifically detects Src in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry
Specifications
| Src | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
| Western Blot, Flow Cytometry, ELISA, Immunocytochemistry | |
| 21H5 | |
| Western Blot 1:500-1:5000, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence 1:20-1:200 | |
| c-src, EC 2.7.10, EC 2.7.10.2, pp60c-src, Rous sarcoma, tyrosine kinase pp60c-src, tyrosine-protein kinase SRC-1, v-src avian sarcoma (Schmidt-Ruppin A-2) viral oncogene homolog, v-src sarcoma (Schmidt-Ruppin A-2) viral oncogene homolog (avian) | |
| A synthesized peptide derived from Human Src [UniProt P12931] | |
| 50 μL | |
| Cancer Stem Cells, Phospho Specific, Protein Kinase, Signal Transduction, Transcription Factors and Regulators, Tyrosine Kinases | |
| 6714 | |
| Store at -20 to -70C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction