missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Src Antibody (21H5), Novus Biologicals™
Rabbit Monoclonal Antibody
€ 355.00 - € 533.00
Specifications
| Antigen | Src |
|---|---|
| Clone | 21H5 |
| Dilution | Western Blot 1:500-1:5000, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence 1:20-1:200 |
| Applications | Western Blot, Flow Cytometry, ELISA, Immunocytochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30035943
|
Novus Biologicals
NBP3-26303-50ul |
50 μL |
€ 355.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30039549
|
Novus Biologicals
NBP3-26303-100ul |
100 μL |
€ 533.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Src Monoclonal antibody specifically detects Src in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, ELISA, ImmunocytochemistrySpecifications
| Src | |
| Western Blot 1:500-1:5000, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence 1:20-1:200 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse | |
| c-src, EC 2.7.10, EC 2.7.10.2, pp60c-src, Rous sarcoma, tyrosine kinase pp60c-src, tyrosine-protein kinase SRC-1, v-src avian sarcoma (Schmidt-Ruppin A-2) viral oncogene homolog, v-src sarcoma (Schmidt-Ruppin A-2) viral oncogene homolog (avian) | |
| A synthesized peptide derived from Human Src [UniProt P12931] | |
| Primary | |
| Store at -20 to -70C. Avoid freeze-thaw cycles. |
| 21H5 | |
| Western Blot, Flow Cytometry, ELISA, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Cancer Stem Cells, Phospho Specific, Protein Kinase, Signal Transduction, Transcription Factors and Regulators, Tyrosine Kinases | |
| PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
| 6714 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title