Learn More
SREBF1 Rabbit, anti-Human, Clone: 1P4, Proteintech
Rabbit Recombinant Monoclonal Antibody
Brand:  Proteintech 82684-3-RR-100UL
 View more versions of this product
Product Code. 18076315
  • € 439.00 / 100µL
Description
The SREBF1 antibody from Proteintech is a rabbit recombinant antibody generated with recombinant protein of human SREBF1. This antibody recognizes human antigen. The SREBF1 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
Specifications
SREBF1
1P4
bHLHd1, SREBF1, SREBP 1, SREBP 1c, SREBP1
100 μL
6720
Store at -20°C. Stable for one year after shipment.
Liquid
Western Blot, ELISA
800 μg/mL
Rabbit
Primary
Human
150 kDa